2'-Aminoacetophenone 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1-(2-Aminophenyl)ethanone, o-Aminoacetophenone

OR28679-100G OR28679-1KG OR28679-500G
APOSOR28679-100GEA 46.2 GBP
APOSOR28679-100G APOSOR28679-1KG APOSOR28679-500G
2'-Aminoacetophenone 98%
2'-Aminoacetophenone
Formula: C₈H₉NO
Boiling Pt: 253…255 °C
Density: 1.118 g/cm³ (25 °C)
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007717
CAS Number: 551-93-9
EINECS: 209-002-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR