α-Chloro-o-xylene 98+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR28627-1KG OR28627-500G OR28627-5KG
APOSOR28627-1KGEA 407 GBP
APOSOR28627-1KG APOSOR28627-500G APOSOR28627-5KG
α-Chloro-o-xylene 98+%
α-Chloro-o-xylene

Intermediate for organic synthesis.

Formula: C₈H₉Cl
Boiling Pt: 197…199 °C
Melting Pt: 2 °C
Density: 1.063 g/cm³ (25 °C)
Flash Pt: 73.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000899
CAS Number: 552-45-4
EINECS: 209-013-8
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR