4-(Benzenesulphonyl)benzaldehyde

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR2728-1G OR2728-5G
APOSOR2728-1GEA 126 GBP
APOSOR2728-1G APOSOR2728-5G
4-(Benzenesulphonyl)benzaldehyde
4-(Benzenesulphonyl)benzaldehyde
Formula: C₁₃H₁₀O₃S
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00444612
CAS Number: 66-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR