Methyl-3,4-diaminobenzoate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3,4-Diamino benzoate, 3,4-Diaminobenzoic acid methyl ester, Methyl 3,4-diaminobenzoate

OR26800-100G OR26800-25G OR26800-5G
APOSOR26800-100GEA 145 GBP
APOSOR26800-100G APOSOR26800-25G APOSOR26800-5G
Methyl-3,4-diaminobenzoate 98%
Methyl 3,4-diaminobenzoate
Formula: C₈H₁₀N₂O₂
Melting Pt: 105…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00017098
CAS Number: 36692-49-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR