2-Chloro-N-(2-{[(fur-2-yl)methyl]thio}ethyl)nicotinamide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR25742-250MG OR25742-50MG
APOSOR25742-250MGEA 83.9 GBP
APOSOR25742-250MG APOSOR25742-50MG
2-Chloro-N-(2-{[(fur-2-yl)methyl]thio}ethyl)nicotinamide
2-Chloro-N-(2-{[(fur-2-yl)methyl]thio}ethyl)nicotinamide
Formula: C₁₃H₁₃ClN₂O₂S
Melting Pt: 42…46 °C
Storage Temperature: Ambient
MDL Number: MFCD01566566
CAS Number: 680211-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR