5-Methylthiazole-2-carbaldehyde

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR2494-1G
APOSOR2494-1GEA 62 GBP
APOSOR2494-1G
5-Methylthiazole-2-carbaldehyde
5-Methylthiazole-2-carbaldehyde
Formula: C5H5NOS
MW: 127.1643 g/mol
Boiling Pt: 80…81 °C/7.5mmHg
Storage Temperature: Ambient
MDL Number: MFCD08060733
CAS Number: 13838-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR