[2-(4-Morpholinyl)phenyl]methanamine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR23385-1G OR23385-5G
APOSOR23385-1GEA 99 GBP
APOSOR23385-1G APOSOR23385-5G
[2-(4-Morpholinyl)phenyl]methanamine 97%
[2-(4-Morpholinyl)phenyl]methanamine
Formula: C₁₁H₁₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD03086183
CAS Number: 204078-48-8
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR