6-Phenoxynicotinic acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR23316-1G
APOSOR23316-1GEA 52.5 GBP
APOSOR23316-1G
6-Phenoxynicotinic acid 98%
6-Phenoxynicotinic acid
Formula: C₁₂H₉NO₃
Melting Pt: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD02682068
CAS Number: 51362-38-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR