Isonicotinic acid 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Pyridine-4-carboxylic acid, 4-Pyridinecarboxylic acid, 4-Picolinic acid

OR23064-500G OR23064-25G OR23064-1KG
APOSOR23064-500GEA 65.1 GBP
APOSOR23064-500G APOSOR23064-25G APOSOR23064-1KG
Isonicotinic acid 99%
Isonicotinic acid
Formula: C₆H₅NO₂
Boiling Pt: 260 °C/15mmHg
Melting Pt: 310…315 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00006429
CAS Number: 55-22-1
EINECS: 200-228-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR