4-Ethynylanisole 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR21951-25G OR21951-5G OR21951-100G
APOSOR21951-25GEA 41 GBP
APOSOR21951-25G APOSOR21951-5G APOSOR21951-100G
4-Ethynylanisole 98%
4-Ethynylanisole
Formula: C₉H₈O
Boiling Pt: 87…91 °C/11mmHg
Melting Pt: 28…29 °C
Density: 1.019 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00168815
CAS Number: 768-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR