2,3-Dimethyl-7-nitroindole 95+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR2112-250MG
APOSOR2112-250MGEA 15.8 GBP
APOSOR2112-250MG
2,3-Dimethyl-7-nitroindole 95+%
2,3-Dimethyl-7-nitroindole
Formula: C₁₀H₁₀N₂O₂
Melting Pt: 164…165 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022710
CAS Number: 41018-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR