2,5-Dibromo-4-picoline

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2,5-Dibromo-4-methylpyridine

OR2087-100G OR2087-25G OR2087-5G
APOSOR2087-100GEA 99.9 GBP
APOSOR2087-100G APOSOR2087-25G APOSOR2087-5G
2,5-Dibromo-4-picoline
2,5-Dibromo-4-picoline
Formula: C₆H₅Br₂N
Melting Pt: 36…42 °C
Storage Temperature: Ambient
MDL Number: MFCD00234955
CAS Number: 3430-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR