3-Amino-4-methylpyridine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR2023-100G
APOSOR2023-100GEA 44.1 GBP
APOSOR2023-100G
3-Amino-4-methylpyridine 98%
3-Amino-4-methylpyridine
Formula: C₆H₈N₂
Boiling Pt: 254 °C
Melting Pt: 102…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00128871
CAS Number: 3430-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR