5-Methylfuran-2-boronic acid 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR2011-1G OR2011-250MG OR2011-5G
APOSOR2011-1GEA 39.9 GBP
APOSOR2011-1G APOSOR2011-250MG APOSOR2011-5G
5-Methylfuran-2-boronic acid 97%
5-Methylfuran-2-boronic acid
Formula: C₅H₇BO₃
Melting Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01114644
CAS Number: 62306-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR