(S,S)-(+)-2,5-Hexanediol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR20022-5G
APOSOR20022-5GEA 70.4 GBP
APOSOR20022-5G
(S,S)-(+)-2,5-Hexanediol
(S,S)-(+)-2,5-Hexanediol
Formula: C₆H₁₄O₂
Boiling Pt: 212…215 °C
Melting Pt: 50…53 °C
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00082583
CAS Number: 34338-96-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR