1-[2-(2-Bromophenoxy)ethyl]piperidine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR200099-1G OR200099-250MG OR200099-500MG OR200099-2.5G
APOSOR200099-1GEA 180 GBP
APOSOR200099-1G APOSOR200099-250MG APOSOR200099-500MG APOSOR200099-2.5G
1-[2-(2-Bromophenoxy)ethyl]piperidine
1-[2-(2-Bromophenoxy)ethyl]piperidine
Formula: C₁₃H₁₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD01308393
CAS Number: 864424-01-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR