1,5-Dimethyl-1H-pyrazole-3-carboxamide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR200066-1G
APOSOR200066-1GEA 94.9 GBP
APOSOR200066-1G
1,5-Dimethyl-1H-pyrazole-3-carboxamide
1,5-Dimethyl-1H-pyrazole-3-carboxamide
Formula: C₆H₉N₃O
Melting Pt: 177…178 °C
Storage Temperature: Ambient
MDL Number: MFCD05664820
CAS Number: 54384-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR