2,6-Dichloronicotinic acid 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2,6-Dichloro-3-pyridinecarboxylic acid, 2,6-Dichloropyridine-3-carboxylic acid

OR1907-100G OR1907-25G OR1907-5G
APOSOR1907-100GEA 77.7 GBP
APOSOR1907-100G APOSOR1907-25G APOSOR1907-5G
2,6-Dichloronicotinic acid 97%
2,6-Dichloronicotinic acid
Formula: C₆H₃Cl₂NO₂
Melting Pt: 140…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00075583
CAS Number: 38496-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR