1-(2-Hydroxyethyl)piperazine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR18310-500G
APOSOR18310-500GEA 21 GBP
APOSOR18310-500G
1-(2-Hydroxyethyl)piperazine 97%
1-(2-Hydroxyethyl)piperazine
Formula: C₆H₁₄N₂O
Boiling Pt: 242…246 °C
Melting Pt: –39 °C
Density: 1.061 g/cm³ (25 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005970
CAS Number: 103-76-4
EINECS: 203-142-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR