3-(Methylamino)propanamide hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR17506-5G OR17506-1G
APOSOR17506-5GEA 401 GBP
APOSOR17506-5G APOSOR17506-1G
3-(Methylamino)propanamide hydrochloride
3-(Methylamino)propanamide hydrochloride
Formula: C₄H₁₁ClN₂O
Density: 1.052 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD11226474
CAS Number: 4874-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR