(R)-3-Methylmorpholine hydrochloride 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR17129-1G
APOSOR17129-1GEA 15.8 GBP
APOSOR17129-1G
(R)-3-Methylmorpholine hydrochloride 97%
(R)-3-Methylmorpholine hydrochloride
Formula: C₅H₁₂ClNO
MW: 137.6100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18382515
CAS Number: 953780-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR