5-Bromo-o-cresol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR16945-25G OR16945-5G OR16945-100G
APOSOR16945-25GEA 27.3 GBP
APOSOR16945-25G APOSOR16945-5G APOSOR16945-100G
5-Bromo-o-cresol
5-Bromo-o-cresol
Formula: C₇H₇BrO
Melting Pt: 75…80 °C
Storage Temperature: Ambient
MDL Number: MFCD08061906
CAS Number: 36138-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR