2-Hydroxy-6-methylbenzoic acid 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2,6-Cresotic acid, 6-Hydroxy-o-toluic acid, 6-Methylsalicylic acid

OR16150-1G OR16150-5G OR16150-10G
APOSOR16150-1GEA 15.8 GBP
APOSOR16150-1G APOSOR16150-5G APOSOR16150-10G
2-Hydroxy-6-methylbenzoic acid 95%
2-Hydroxy-6-methylbenzoic acid
Formula: C₈H₈O₃
Melting Pt: 170…171 °C
Storage Temperature: Ambient
MDL Number: MFCD01194284
CAS Number: 567-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR