3-Boc-aminopyrrolidine 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3-(BOC-amino)pyrrolidine, 2-Methyl-2-propanyl 3-pyrrolidinylcarbamate, 1,1-Dimethylethyl3-pyrrolidinylcarbamate, 3-Boc-Aminopyrrolidine

OR15678-5G OR15678-1G OR15678-25G
APOSOR15678-5GEA 52.5 GBP
APOSOR15678-5G APOSOR15678-1G APOSOR15678-25G
3-Boc-aminopyrrolidine 95%
3-Boc-aminopyrrolidine
Formula: C₉H₁₈N₂O₂
Melting Pt: 63…68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00059040
CAS Number: 99724-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR