Ethyl-2-isopropylthiazole-4-carboxylate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Ethyl 2-isopropylthiazole-4-carboxylate

OR15537-1G
APOSOR15537-1GEA 27.3 GBP
APOSOR15537-1G
Ethyl-2-isopropylthiazole-4-carboxylate
Ethyl-2-isopropylthiazole-4-carboxylate
Formula: C₉H₁₃NO₂S
MW: 199.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09025787
CAS Number: 133047-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR