Ethyl-2-aminonicotinate 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR15285-25G OR15285-5G
APOSOR15285-25GEA 52.5 GBP
APOSOR15285-25G APOSOR15285-5G
Ethyl-2-aminonicotinate 98%
Ethyl 2-aminonicotinate
Formula: C₈H₁₀N₂O₂
Boiling Pt: 266.75 °C
Melting Pt: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD01569452
CAS Number: 13362-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR