2,5-Dichloroanisole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR15004-25G OR15004-5G
APOSOR15004-25GEA 37.8 GBP
APOSOR15004-25G APOSOR15004-5G
2,5-Dichloroanisole
2,5-Dichloroanisole
Formula: C₇H₆Cl₂O
Boiling Pt: 128…132 °C/3.5mmHg
Density: 1.33 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00061123
CAS Number: 1984-58-3
EINECS: 217-852-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR