5-Chloro-2-methylanisole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR15003-5G
APOSOR15003-5GEA 17.9 GBP
APOSOR15003-5G
5-Chloro-2-methylanisole
5-Chloro-2-methylanisole
Formula: C₈H₉ClO
Storage Temperature: Ambient
MDL Number: MFCD07368289
CAS Number: 40794-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR