1-Methyl-5-[(methylamino)methyl]indoline 90%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR14864-1G OR14864-250MG
APOSOR14864-1GEA 258 GBP
APOSOR14864-1G APOSOR14864-250MG
1-Methyl-5-[(methylamino)methyl]indoline 90%
1-Methyl-5-[(methylamino)methyl]indoline
Formula: C₁₁H₁₆N₂
Storage Temperature: Refrigerator
MDL Number: MFCD09817499
CAS Number: 906352-81-6
UN: 2922
ADR: 8+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR