Fmoc-3-methyl-L-phenylalanine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3-Methyl-L-phenylalanine, N-FMOC protected, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(m-tolyl)propanoic acid, Fmoc-Phe(3-Me)-OH

OR14570-25G OR14570-5G
APOSOR14570-25GEA 393 GBP
APOSOR14570-25G APOSOR14570-5G
Fmoc-3-methyl-L-phenylalanine 97%
Fmoc-3-methyl-L-phenylalanine
Formula: C₂₅H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD01863050
CAS Number: 211637-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR