2-Amino-5-hydroxymethylpyridine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR14503-1G OR14503-250MG
APOSOR14503-1GEA 47.3 GBP
APOSOR14503-1G APOSOR14503-250MG
2-Amino-5-hydroxymethylpyridine 97%
2-Amino-5-hydroxymethylpyridine
Formula: C₆H₈N₂O
Melting Pt: 113…120 °C
Storage Temperature: Ambient
MDL Number: MFCD06659089
CAS Number: 113293-71-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR