Methyl-3,5-dichloro-4-methoxybenzenecarboxylate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR14469-10G OR14469-1G OR14469-5G
APOSOR14469-10GEA 647 GBP
APOSOR14469-10G APOSOR14469-1G APOSOR14469-5G
Methyl-3,5-dichloro-4-methoxybenzenecarboxylate
Methyl-3,5-dichloro-4-methoxybenzenecarboxylate
Formula: C₉H₈Cl₂O₃
Melting Pt: 74…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00017189
CAS Number: 24295-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR