3-Ethyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3-Ethyl-2-thioxo-2,3-dihydroquinazolin-4(1H)-one

OR14408-1G
APOSOR14408-1GEA 21 GBP
APOSOR14408-1G
3-Ethyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
3-Ethyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Formula: C₁₀H₁₀N₂OS
MW: 206.27 g/mol
Melting Pt: 256…258 °C
Storage Temperature: Ambient
MDL Number: MFCD00126093
CAS Number: 13906-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR