2-Amino-3-hydroxy-3-(4-methoxyphenyl)propanoic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR13802-250MG
APOSOR13802-250MGEA 168 GBP
APOSOR13802-250MG
2-Amino-3-hydroxy-3-(4-methoxyphenyl)propanoic acid
2-Amino-3-hydroxy-3-(4-methoxyphenyl)propanoic acid
Formula: C₁₀H₁₃NO₄
Melting Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD09998363
CAS Number: 50897-30-8
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR