tert-Butyl hydrogen malonate 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3-Tert-butoxy-3-oxopropanoic acid, mono-tert-Butyl malonate

OR13710-100G OR13710-25G OR13710-5G
APOSOR13710-100GEA 49.4 GBP
APOSOR13710-100G APOSOR13710-25G APOSOR13710-5G
tert-Butyl hydrogen malonate 95%
tert-Butyl hydrogen malonate
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 90 °C/2mmHg
Melting Pt: 19…20 °C
Density: 1.04 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00191886
CAS Number: 40052-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR