Pyridinium chloride 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Pyridine hydrochloride, Pyridinehydrochloride, Pyridine HCl

OR13636-100G OR13636-25G
APOSOR13636-100GEA 26.3 GBP
APOSOR13636-100G APOSOR13636-25G
Pyridinium chloride 95%
Pyridinium chloride
Formula: C₅H₆ClN
Boiling Pt: 222…224 °C
Melting Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00012802
CAS Number: 628-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR