5-Oxo-1-(1-phenylethyl)pyrrolidine-3-carboxylic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR13465-1G
APOSOR13465-1GEA 48.3 GBP
APOSOR13465-1G
5-Oxo-1-(1-phenylethyl)pyrrolidine-3-carboxylic acid
5-Oxo-1-(1-phenylethyl)pyrrolidine-3-carboxylic acid
Formula: C₁₃H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD02625781
CAS Number: 96449-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR