Benzotriazole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR13222-1KG OR13222-250G OR13222-500G
APOSOR13222-1KGEA 44.1 GBP
APOSOR13222-1KG APOSOR13222-250G APOSOR13222-500G
Benzotriazole
Benzotriazole
Formula: C₆H₅N₃
Boiling Pt: 204 °C/15mmHg
Melting Pt: 96…99 °C
Density: 1.36 g/cm³ (25 °C)
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00005699
CAS Number: 95-14-7
EINECS: 202-394-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR