Methyl isonicotinate 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR13188-100G OR13188-1G
APOSOR13188-100GEA 57.8 GBP
APOSOR13188-100G APOSOR13188-1G
Methyl isonicotinate 97%
Methyl isonicotinate
Formula: C₇H₇NO₂
Boiling Pt: 207…209 °C
Melting Pt: 8…8.5 °C
Density: 1.161 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006427
CAS Number: 2459-09-8
EINECS: 219-546-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR