Ethyl-2-methylthiazole-4-carboxylate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR1301-10G OR1301-1G OR1301-5G
APOSOR1301-10GEA 25.2 GBP
APOSOR1301-10G APOSOR1301-1G APOSOR1301-5G
Ethyl-2-methylthiazole-4-carboxylate
Ethyl 2-methylthiazole-4-carboxylate
Formula: C₇H₉NO₂S
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00156148
CAS Number: 6436-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR