4-(Phenylsulphonyl)piperidine hydrochloride 95+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Phenyl piperidin-4-yl sulphone hydrochloride, 4-Benzenesulphonylpiperidine Hydrochloride

OR12886-1G OR12886-5G
APOSOR12886-1GEA 185 GBP
APOSOR12886-1G APOSOR12886-5G
4-(Phenylsulphonyl)piperidine hydrochloride 95+%
4-(Phenylsulphonyl)piperidine hydrochloride
Formula: C₁₁H₁₅NO₂S·ClH
Storage Temperature: Ambient
MDL Number: MFCD03840151
CAS Number: 1172500-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR