5-Bromoindole 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 5-Bromo-1H-indole

OR1278-100G OR1278-500G
APOSOR1278-100GEA 23.1 GBP
APOSOR1278-100G APOSOR1278-500G
5-Bromoindole 99%
5-Bromoindole
Formula: C₈H₆BrN
Melting Pt: 90…92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005670
CAS Number: 10075-50-0
EINECS: 233-208-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR