N-Boc-1,3-propanediamine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: N-(3-Aminopropyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(3-Aminopropyl)carbamate, N-Boc-1,3-propanediamine, tert-Butyl (3-aminopropyl)carbamate, N-1-Boc-1,3-diaminopropane·HCl, N-(tert-Butoxycarbonyl)-1,3-propanediamine, N-Boc-1,3-diaminopropane

OR1269-25G OR1269-5G
APOSOR1269-25GEA 31.5 GBP
APOSOR1269-25G APOSOR1269-5G
N-Boc-1,3-propanediamine
N-Boc-1,3-propanediamine
Formula: C₈H₁₈N₂O₂
Boiling Pt: 203 °C
Melting Pt: 22 °C
Density: 0.998 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00210021
CAS Number: 75178-96-0
UN: 3259
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR