5-(Bromoacetyl)-3-phenylisoxazole 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR1268-1G OR1268-250MG OR1268-5G
APOSOR1268-1GEA 52.5 GBP
APOSOR1268-1G APOSOR1268-250MG APOSOR1268-5G
5-(Bromoacetyl)-3-phenylisoxazole 95%
5-(Bromoacetyl)-3-phenylisoxazole
Formula: C₁₁H₈BrNO₂
Melting Pt: 109…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00173898
CAS Number: 14731-14-7
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR