Homophthalic anhydride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR12643-100G OR12643-25G OR12643-5G
APOSOR12643-100GEA 206 GBP
APOSOR12643-100G APOSOR12643-25G APOSOR12643-5G
Homophthalic anhydride
Homophthalic anhydride
Formula: C₉H₆O₃
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00006894
CAS Number: 703-59-3
EINECS: 211-873-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR