Methyl-3-amino-5-hydroxybenzoate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR12411-1G OR12411-25G OR12411-5G
APOSOR12411-1GEA 126 GBP
APOSOR12411-1G APOSOR12411-25G APOSOR12411-5G
Methyl-3-amino-5-hydroxybenzoate
Methyl 3-amino-5-hydroxybenzoate
Formula: C₈H₉NO₃
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD09263220
CAS Number: 67973-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR