2-(2-Methylimidazol-1-yl)phenylamine 95+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR12232-1G
APOSOR12232-1GEA 141 GBP
APOSOR12232-1G
2-(2-Methylimidazol-1-yl)phenylamine 95+%
2-(2-Methylimidazol-1-yl)phenylamine
Formula: C₁₀H₁₁N₃
Melting Pt: 132.5…134.5 °C
Storage Temperature: Ambient
MDL Number: MFCD08699292
CAS Number: 26286-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR