Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Methyl 3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate

OR12149-1G
APOSOR12149-1GEA 16.8 GBP
APOSOR12149-1G
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate
Formula: C₁₀H₉NO₄
MW: 207.185 g/mol
Melting Pt: 183…185 °C
Storage Temperature: Ambient
MDL Number: MFCD03036646
CAS Number: 202195-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR