6-Methyl-1H-indazol-5-amine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR12141-1G OR12141-250MG
APOSOR12141-1GEA 79.8 GBP
APOSOR12141-1G APOSOR12141-250MG
6-Methyl-1H-indazol-5-amine
6-Methyl-1H-indazol-5-amine
Formula: C₈H₉N₃
Melting Pt: 215…217 °C
Storage Temperature: Freezer
MDL Number: MFCD03305372
CAS Number: 81115-45-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR