2-Chloro-4-methylpyrimidine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR12067-5G OR12067-100G OR12067-25G
APOSOR12067-5GEA 15.8 GBP
APOSOR12067-5G APOSOR12067-100G APOSOR12067-25G
2-Chloro-4-methylpyrimidine
2-Chloro-4-methylpyrimidine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 93…94 °C/17mmHg
Melting Pt: 45…53 °C
Flash Pt: 98 °C
Storage Temperature: Freezer
MDL Number: MFCD00054434
CAS Number: 13036-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR